Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PIGF Partial ORF (NP_775097.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89956903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-903 10 ug
89-956-904 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-956-903 Supplier Abnova™ Supplier No. H00005281Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor, a glycolipid containing three mannose molecules in its core backbone, is found on many blood cells where it serves to anchor proteins to the cell surface. The encoded protein and another GPI synthesis protein, PIGO, function in the transfer of ethanolaminephosphate to the third mannose in GPI. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGFVTAVNLVLYLVVKPNTSSKRSSLSHKVTGFLKCCIYFLMSCFSFHVIFVL

Specifications

Accession Number NP_775097.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5281
Molecular Weight (g/mol) 36.63kDa
Name PIGF (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 10 ug
Immunogen MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGFVTAVNLVLYLVVKPNTSSKRSSLSHKVTGFLKCCIYFLMSCFSFHVIFVL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC32646/MGC33136
Common Name PIGF
Gene Symbol PIGF
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.