Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PIM1 (P11309) Full-length Recombinant Protein with GST expressed in Sf9 cells

Catalog No. 89956984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-984 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-984 Supplier Abnova™ Supplier No. P4225

Used for Func, SDS-PAGE

The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer (Dhanasekaran et al., 2001 [PubMed 11518967]).[supplied by OMIM]

Specifications

Accession Number P11309
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5292
Molecular Weight (g/mol) 62kDa
Name PIM1 (Human) Recombinant Protein
Quality Control Testing 1 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PIM
Common Name PIM1
Gene Symbol PIM1
Biological Activity 316nmol phosphate incorporated into the S6K substrate peptide per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay).
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.