Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PISD Full-length ORF (CAK54571.1, 1 a.a. - 409 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-048 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-048 Supplier Abnova™ Supplier No. H00023761P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Phosphatidylserine decarboxylases (PSDs; EC 4.1.1.65) catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits (Schuiki and Daum, 2009 [PubMed 19165886]).[supplied by OMIM]

Sequence: MATSVGHRCLGLLHGVAPWRSSLHPCEITALSQSLQPLRKLPFRAFRTDARKIHTAPARTMFLLRPLPILLVTGGGYAGYRQYEKYRERELEKLGLEIPPKLAGHWEVALYKSVPTRLLSRAWGRLNQVELPHWLRRPVYSLYIWTFGVNMKEAAVEDLHHYRNLSEFFRRKLKPQARPVCGLHSVISPSDGRILNFGQVKNCEVEQVKGVTYSLESFLGPRMCTEDLPFPPAASCDSFKNQLVTREGNELYHCVIYLAPGDYHCFHSPTDWTVSHRRHFPGSLMSVNPGMARWIKELFCHNERVVLTGDWKHGFFSLTAVGATNVGSIRIYFDRDLHTNSPRHSKGSYNDFSFVTHTNREGVPMRKGEHLGEFNLGSTIVLIFEAPKDFNFQLKTGQKIRFGEALGSL

Specifications

Accession Number CAK54571.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23761
Molecular Weight (g/mol) 71.39kDa
Name PISD (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MATSVGHRCLGLLHGVAPWRSSLHPCEITALSQSLQPLRKLPFRAFRTDARKIHTAPARTMFLLRPLPILLVTGGGYAGYRQYEKYRERELEKLGLEIPPKLAGHWEVALYKSVPTRLLSRAWGRLNQVELPHWLRRPVYSLYIWTFGVNMKEAAVEDLHHYRNLSEFFRRKLKPQARPVCGLHSVISPSDGRILNFGQVKNCEVEQVKGVTYSLESFLGPRMCTEDLPFPPAASCDSFKNQLVTREGNELYHCVIYLAPGDYHCFHSPTDWTVSHRRHFPGSLMSVNPGMARWIKELFCHNERVVLTGDWKHGFFSLTAVGATNVGSIRIYFDRDLHTNSPRHSKGSYNDFSFVTHTNREGVPMRKGEHLGEFNLGSTIVLIFEAPKDFNFQLKTGQKIRFGEALGSL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DJ858B16/DKFZp566G2246/PSDC/PSSC/dJ858B16.2
Common Name PISD
Gene Symbol PISD
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.