Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PITX1 Full-length ORF (NP_002644.3, 1 a.a. - 314 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957067 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-067 10 ug
89-957-068 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-067 Supplier Abnova™ Supplier No. H00005307P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family are involved in organ development and left-right asymmetry. This protein acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. [provided by RefSeq]

Sequence: MDAFKGGMSLERLPEGLRPPPPPPHDMGPAFHLARPADPREPLENSASESSDTELPEKERGGEPKGPEDSGAGGTGCGGADDPAKKKKQRRQRTHFTSQQLQELEATFQRNRYPDMSMREEIAVWTNLTEPRVRVWFKNRRAKWRKRERNQQLDLCKGGYVPQFSGLVQPYEDVYAAGYSYNNWAAKSLAPAPLSTKSFTFFNSMSPLSSQSMFSAPSSISSMTMPSSMGPGAVPGMPNSGLNNINNLTGSSLNSAMSPGACPYGTPASPYSVYRDTCNSSLASLRLKSKQHSSFGYGALQGPASGLNACQYNS

Specifications

Accession Number NP_002644.3
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5307
Molecular Weight (g/mol) 60.5kDa
Name PITX1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MDAFKGGMSLERLPEGLRPPPPPPHDMGPAFHLARPADPREPLENSASESSDTELPEKERGGEPKGPEDSGAGGTGCGGADDPAKKKKQRRQRTHFTSQQLQELEATFQRNRYPDMSMREEIAVWTNLTEPRVRVWFKNRRAKWRKRERNQQLDLCKGGYVPQFSGLVQPYEDVYAAGYSYNNWAAKSLAPAPLSTKSFTFFNSMSPLSSQSMFSAPSSISSMTMPSSMGPGAVPGMPNSGLNNINNLTGSSLNSAMSPGACPYGTPASPYSVYRDTCNSSLASLRLKSKQHSSFGYGALQGPASGLNACQYNS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BFT/POTX/PTX1
Common Name PITX1
Gene Symbol PITX1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.