Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLA2G7/PAF-AH/Lp-PLA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5106SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5106SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5106SP Supplier R&D Systems Supplier No. AF5106SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PLA2G7/PAF-AH/Lp-PLA2 Polyclonal specifically detects PLA2G7/PAF-AH/Lp-PLA2 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.

Specifications

Antigen PLA2G7/PAF-AH/Lp-PLA2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH38452
Gene Alias 2-acetyl-1-alkylglycerophosphocholine esterase, EC 3.1.1, EC 3.1.1.47,1-alkyl-2-acetylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), LDL-PLA2, lipoprotein-associated phospholipase A2, LpPLA2, LP-PLA2, PAF acetylhydrolase, PAF-AH, PAFAHPAF 2-acylhydrolase, phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma), platelet-activating factor acetylhydrolase
Gene Symbols PLA2G7
Host Species Goat
Immunogen E. coli-derived recombinant human PLA2G7/PAF-AH/Lp-PLA2 Met33-Asn441 Accession # AAH38452
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7941
Test Specificity Detects human PLA2G7/PAF-AH/Lp-PLA2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) PLA2G-2A, recombinant mouse PLA2G-2A, and rhPLA2G-1B is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.