Learn More
Description
- Also known as: PLA2R; PLA2-R; PLA2IR; CLEC13C; PLA2G1R
- PLA2R1 is a phospholipase A2 receptor.
- It exists as both a transmembrane form and a soluble form.
- The transmembrane receptor may play a role in clearance of phospholipase A2, thereby inhibiting its action.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hPLA2R1 |
| Accession Number | NP_031392 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 4405 bp |
| Gene ID (Entrez) | 22925 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
