Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Placental Lactogen/CSH1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5757SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5757SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5757SP Supplier R&D Systems Supplier No. MAB5757SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Placental Lactogen/CSH1 Monoclonal specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Placental Lactogen/CSH1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 658230
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01243
Gene Alias Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH1, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, PL, Placental lactogen, PLchoriomammotropin
Gene Symbols CSH1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Placental Lactogen/CSH1 Val27-Phe217 Accession # P01243
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1442
Test Specificity Detects human Placental Lactogen/CSH1 in direct ELISAs and Western blot. In direct ELISAs, no cross-reactivity with recombinant human Growth Hormone is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.