Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PLAGL2 Synthetic Peptide

Catalog No. PEP1499
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1499 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1499 Supplier Invitrogen™ Supplier No. PEP1499
Add to Cart
Add to Cart

Synthetic Peptide

PLAGL2 Synthetic Peptide, PEP-1499, from Invitrogen.

The PLAG (Pleiomorphic adenoma gene) family of zinc finger proteins include PLAG1, ZAC1 and PLAG-like 2 (PLAGL2). PLAG1 is the target gene for the pleiomorphic adenomas of the salivary gland. ZAC1 (PLAGL1) concomitantly controls apoptosis and cell cycle arrest through separate pathways. ZAC1 also acts as a transcriptional cofactor for nuclear receptors. PLAGL2 belongs to the krueppel C2H2-type zinc-finger protein family and contains 6 C2H2-type zinc fingers. It functions as a positive regulator of transcription and localizes to the nucleus. PLAGL2 is ubiquitously expressed with particular abundance in spleen, lung and testis, where it may be involved in cell cycle arrest and apoptosis of tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5326
Species Human
Purification Method purified
Accession Number Q9UPG8
Common Name Human PLAGL2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol PLAGL2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.