Learn More
Innovative Research Inc Human Plasmin Purified 1mg
Supplier:  Innovative Research Inc IHUPLM1MG
Encompass_Preferred
Human Plasmin Purified from Innovative Research has been activated from glu-Plasminogen using Urokinase (uPA) coupled resin. This is a frozen liquid buffered in 50mM Hepes/50mM Sodium Acetate/pH 8.5/50% Glycerol, with a concentration of 1mg/ml. Complete activation was observed by S-2251 chromogenic substrate.
Catalog No. 50-203-6507
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.