Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Plasminogen Protein

Catalog No. 1939SE200
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1939SE200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1939SE200 Supplier R&D Systems™ Supplier No. 1939SE200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, SUC-AFK-AMC. The specific activity, measured under the described conditions, is >500 pmol/min/μg.

Specifications

For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 5340
Molecular Weight (g/mol) M.W. (Observed): 100-106 kDa doublet, reducing conditions
Name Plasminogen
Quantity 200 μg
Source Plasminogen protein The human plasma used for the isolation of this product was certified by the supplier to be HIV-1 and HBsAg negative (at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions.)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol PLG
Conjugate Unconjugated
Reconstitution Reconstitute at 1 mg/mL in sterile 25 mM Tris and 150 mM NaCl, pH 8.0.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Plasminogen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.