Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Pleiotrophin Recombinant Protein, PeproTech®

Catalog No. 4501550UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-15-50UG 50 μg
45015100UG 100 μg
450-15-1MG 1 mg
450-15-20UG 20 μg
45015250UG 250 μg
45015500UG 500 μg
450-15-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 450-15-50UG Supplier Gibco™ Supplier No. 4501550UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Pleiotrophin is a 15.4 kDa protein containing 136 amino acid residues and five intra-molecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Pleiotrophin (PTN) is an 18 kDa secreted heparin and chondroitin-binding protein that is a regulatory cytokine critical to the early development of blood vessels and neurons. High PTN serum levels are associated with a variety of solid tumors. It was shown that patients with multiple myeloma (MM) also have elevated serum levels of this protein. The expression of this protein is very limited in normal adult tissues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21246
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 5764
Molecular Weight (g/mol) 15.4 kDa
Name Human Pleiotrophin
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN
Common Name Pleiotrophin
Gene Symbol PTN
Conjugate Unconjugated
Recombinant Recombinant
Sequence GKKEKPEKKV KKSDCGEWQW SVCVPTSGDC GLGTREGTRT GAECKQTMKT QRCKIPCNWK KQFGAECKYQ FQAWGECDLN TALKTRTGSL KRALHNAECQ KTVTISKPCG KLTKPKPQAE SKKKKKEGKK QEKMLD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.