Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PLEKHM1 Synthetic Peptide

Catalog No. PEP0971
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0971 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0971 Supplier Invitrogen™ Supplier No. PEP0971
Add to Cart
Add to Cart

Synthetic Peptide

PLEKHM1 Synthetic Peptide, PEP-0971, from Invitrogen.

PLEKHM1 is a member of the M family of Pleckstrin homolog domain-containing proteins, a group of proteins containing a RUN domain, two pleckstrin homology domains, and a cysteine-rich domain. It was identified through segregation analysis as a cause of osteopetrosis in humans. PLEKHM1 co-localizes with Rab7 to late endosomal/lysosomal vesicles in HEK293 and osteoclast-like cells, with this co-localization dependent on the prenylation of Rab7. Monocytes from a patient homozygous for a mutated form of PLEKHM1differentiated into osteoclasts normally, but failed to form ruffled borders and showed little evidence of bone resorption when cultured on dentine discs. Another mutation of PLEKHM1 impaired vesicular acidification and increased TRACP secretion in osteoclasts, suggesting that PLEKHM1 has critical roles in endosomal maturation and may be important in osteoclast-osteoblast cross-talk.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9842
Species Human
Purification Method purified
Accession Number Q9Y4G2
Common Name Human PLEKHM1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol PLEKHM1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.