Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PLEKHM3 Synthetic Peptide

Catalog No. PEP0965
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0965 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0965 Supplier Invitrogen™ Supplier No. PEP0965
Add to Cart
Add to Cart

Synthetic Peptide

PLEKHM3 Synthetic Peptide, PEP-0965, from Invitrogen.

PLEKHM3, also known as DAPR, is a member of the M family of Pleckstrin homology domain-containing proteins. PLEKHM3 was initially identified through chromatin immunoprecipitation and CpG microarray analysis examining proteins regulated by myocyte-enhancing factor 2. In C2C12 myoblast cells, PLEKHM3 binds to the PI3K signaling member protein kinase B in the cytosol prior to differentiation into myotubes. Following the initiation of differentiation, PLEKHM3 was also found in membrane fractions. Knockdown of PLEKHM3 expression by RNAi resulted in the inhibition of myotube formation, suggesting that PLEKHM3 is a key component required by myoblasts for orchestrating their differentiation during myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 389072
Species Human
Purification Method purified
Accession Number Q6ZWE6
Common Name Human PLEKHM3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol PLEKHM3
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.