Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Plexin A1 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB6536SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6536SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6536SP Supplier R&D Systems Supplier No. MAB6536SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plexin A1 Monoclonal specifically detects Plexin A1 in Human samples. It is validated for Immunocytochemistry∣Western Blot.

Specifications

Antigen Plexin A1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 659807
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UIW2
Gene Alias NOV, NOVP, plexin A1, PLEXIN-A1, PLXN1, PLXNA1
Gene Symbols PLXNA1
Host Species Mouse
Immunogen E. coli-derived recombinant human Plexin A1 Asn986-Leu1152 Accession # Q9UIW2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5361
Test Specificity Detects human Plexin A1 in direct ELISAs. In Western blots, no cross-reactivity with recombinant human Plexin A4, B1, B2, B3, C1, D1, recombinant mouse Plexin A1, A2, or A3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.