Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human PlGF-2 Recombinant Protein, PeproTech®

Catalog No. 10056A500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10056A500UG 500 μg
10056A100UG 100 μg
100-56A-1MG 1 mg
10056A250UG 250 μg
10056A25UG 25 μg
100-56A-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10056A500UG Supplier Gibco™ Supplier No. 10056A500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human PlGF-2 is a 34.6 kDa disulfide-linked homodimeric protein of two 152 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

PlGF-2 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-2 is expressed in umbilical vein endothelial cells and placenta. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration. PlGF-2 also signals through Neuropilin (NP-1), and can bind with high affinity to heparin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49763
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 5228
Molecular Weight (g/mol) 34.6 kDa
Name Human PlGF-2
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name PlGF-2
Gene Symbol PGF
Biological Activity Determined by its ability to chemoattract human THP-1 cells of using a concentration range of 5.0-50.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPAVPPQQWA LSAGNGSSEV EVVPFQEVWG RSYCRALERL VDVVSEYPSE VEHMFSPSCV SLLRCTGCCG DENLHCVPVE TANVTMQLLK IRSGDRPSYV ELTFSQHVRC ECRPLREKMK PERRRPKGRG KRRREKQRPT DCHLCGDAVP RR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.