Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human PlGF-3 Recombinant Protein, PeproTech®

Catalog No. 100575UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-57-5UG 5 μg
10057100UG 100 μg
100-57-1MG 1 mg
10057250UG 250 μg
100-57-25UG 25 μg
10057500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 100-57-5UG Supplier Gibco™ Supplier No. 100575UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human PlGF-3 is a 45.7 kDa disulfide-linked homodimeric protein of two 203 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49763
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 5228
Molecular Weight (g/mol) 45.7 kDa
Name Human PlGF-3
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760
Common Name PGF
Gene Symbol PGF
Biological Activity Determined by its ability to chemoattract human monocytes using a concentration range of 5.0-50.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPAVPPQQWA LSAGNGSSEV EVVPFQEVWG RSYCRALERL VDVVSEYPSE VEHMFSPSCV SLLRCTGCCG DENLHCVPVE TANVTMQLLK IRSGDRPSYV ELTFSQHVRC ECRHSPGRQS PDMPGDFRAD APSFLPPRRS LPMLFRMEWG CALTGSQSAV WPSSPVPEEI PRMHPGRNGK KQQRKPLREK MKPERCGDAV PRR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.