Learn More
ACROBiosystems Human PLGF / PGF (19-149) Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems PGFH525620UG
Encompass_Preferred
Recombinant Protein;20UG;Human PLGF (19-149), Fc Tag (PGF-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.2 kDa.
Catalog No. 50-205-5062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.