Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human PlGF Quantikine HS ELISA Kit
SDP

Catalog No. HSPG00 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
HSPG00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HSPG00 Supplier R&D Systems™ Supplier No. HSPG00
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 3.5 hour solid phase ELISA designed to measure PlGF levels in serum and plasma.

The Human PlGF Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PLGF in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).

Specifications

Assay Range 3.13 - 200pg/mL
Assay Sensitivity 0.326pg/mL
Conjugate HRP
Sample Type Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human PlGF
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 5228
Sample Volume Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.