Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PLOD1 Full-length ORF (NP_000293.2, 1 a.a. - 727 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957372 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-372 10 ug
89-957-373 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-372 Supplier Abnova™ Supplier No. H00005351P01S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

Lysyl hydroxylase is a membrane-bound homodimeric protein localized to the cisternae of the endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VI have deficiencies in lysyl hydroxylase activity. [provided by RefSeq]

Sequence: MRPLLLLALLGWLLLAEAKGDAKPEDNLLVLTVATKETEGFRRFKRSAQFFNYKIQALGLGEDWNVEKGTSAGGGQKVRLLKKALEKHADKEDLVILFADSYDVLFASGPRELLKKFRQARSQVVFSAEELIYPDRRLETKYPVVSDGKRFLGSGGFIGYAPNLSKLVAEWEGQDSDSDQLFYTKIFLDPEKREQINITLDHRCRIFQNLDGALDEVVLKFEMGHVRARNLAYDTLPVLIHGNGPTKLQLNYLGNYIPRFWTFETGCTVCDEGLRSLKGIGDEALPTVLVGVFIEQPTPFVSLFFQRLLRLHYPQKHMRLFIHNHEQHHKAQVEEFLAQHGSEYQSVKLVGPEVRMANADARNMGADLCRQDRSCTYYFSVDADVALTEPNSLRLLIQQNKNVIAPLMTRHGRLWSNFWGALSADGYYARSEDYVDIVQGRRVGVWNVPYISNIYLIKGSALRGELQSSDLFHHSKLDPDMAFCANIRQQDVFMFLTNRHTLGHLLSLDSYRTTHLHNDLWEVFSNPEDWKEKYIHQNYTKALAGKLVETPCPDVYWFPIFTEVACDELVEEMEHFGQWSLGNNKDNRIQGGYENVPTIDIHMNQIGFEREWHKFLLEYIAPMTEKLYPGYYTRAQFDLAFVVRYKPDEQPSLMPHHDASTFTINIALNRVGVDYEGGGCRFLRYNCSIRAPRKGWTLMHPGRLTHYHEGLPTTRGTRYIAVSFVDP

Specifications

Accession Number NP_000293.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5351
Molecular Weight (g/mol) 109.9kDa
Name PLOD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MRPLLLLALLGWLLLAEAKGDAKPEDNLLVLTVATKETEGFRRFKRSAQFFNYKIQALGLGEDWNVEKGTSAGGGQKVRLLKKALEKHADKEDLVILFADSYDVLFASGPRELLKKFRQARSQVVFSAEELIYPDRRLETKYPVVSDGKRFLGSGGFIGYAPNLSKLVAEWEGQDSDSDQLFYTKIFLDPEKREQINITLDHRCRIFQNLDGALDEVVLKFEMGHVRARNLAYDTLPVLIHGNGPTKLQLNYLGNYIPRFWTFETGCTVCDEGLRSLKGIGDEALPTVLVGVFIEQPTPFVSLFFQRLLRLHYPQKHMRLFIHNHEQHHKAQVEEFLAQHGSEYQSVKLVGPEVRMANADARNMGADLCRQDRSCTYYFSVDADVALTEPNSLRLLIQQNKNVIAPLMTRHGRLWSNFWGALSADGYYARSEDYVDIVQGRRVGVWNVPYISNIYLIKGSALRGELQSSDLFHHSKLDPDMAFCANIRQQDVFMFLTNRHTLGHLLSLDSYRTTHLHNDLWEVFSNPEDWKEKYIHQNYTKALAGKLVETPCPDVYWFPIFTEVACDELVEEMEHFGQWSLGNNKDNRIQGGYENVPTIDIHMNQIGFEREWHKFLLEYIAPMTEKLYPGYYTRAQFDLAFVVRYKPDEQPSLMPHHDASTFTINIALNRVGVDYEGGGCRFLRYNCSIRAPRKGWTLMHPGRLTHYHEGLPTTRGTRYIAVSFVDP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ42041/LH/LH1/LLH/PLOD
Common Name PLOD1
Gene Symbol PLOD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.