Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLUNC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB18971100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB18971100 100 μg
MAB18971SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB18971100 Supplier R&D Systems Supplier No. MAB18971100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLUNC Monoclonal specifically detects PLUNC in Human samples. It is validated for ELISA.

Specifications

Antigen PLUNC
Applications ELISA
Classification Monoclonal
Clone 251511R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9NP55
Gene Alias bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl
Host Species Mouse
Immunogen E. coli-derived recombinant human PLUNC, Gln20-Val256, Accession No. Q9NP55
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 51297.0
Test Specificity Detects human PLUNC in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.