Learn More
Description
- Verifies the pluripotency of your starting population
- Only takes 5 days
- Reduces experimental variation
Specifications
Specifications
| Cross Reactivity | Human |
| Includes | Differentiation Base Media Supplement (50X); Ectoderm, Mesoderm, and Endoderm Differentiation Supplements; Ectoderm Marker: Goat Anti-Human Otx2 Antigen Affinity-purified Polyclonal Antibody; Mesoderm Marker: Goat Anti-Human Brachyury Antigen Affinity-purified Polyclonal AntibodyV; Endoderm Marker: Goat Anti-Human SOX17 Antigen Affinity-purified Polyclonal Antibody; The quantity of each component in this kit is sufficient to make 200 mL of media for differentiation |
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Form | Liquid |
| Product Line | R&D Systems |
| Product Type | Human Pluripotent Stem Cell Functional Identification Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
