Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PMP22 Full-length ORF (NP_000295.1, 1 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957433 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-433 25 ug
89-957-432 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-433 Supplier Abnova™ Supplier No. H00005376P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an integral membrane protein that is a major component of myelin in the peripheral nervous system. Various mutations of this gene are causes of Charcot-Marie-Tooth disease Type IA, Dejerine-Sottas syndrome, and hereditary neuropathy with liability to pressure palsies. Alternative splicing of this gene results in three transcript variants that encode the same protein. [provided by RefSeq]

Sequence: MLLLLLSIIVLHVAVLVLLFVSTIVSQWIVGNGHATDLWQNCSTSSSGNVHHCFSSSPNEWLQSVQATMILSIIFSILSLFLFFCQLFTLTKGGRFYITGIFQILAGLCVMSAAAIYTVRHPEWHLNSDYSYGFAYILAWVAFPLALLSGVIYVILRKRE

Specifications

Accession Number NP_000295.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5376
Molecular Weight (g/mol) 44.3kDa
Name PMP22 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MLLLLLSIIVLHVAVLVLLFVSTIVSQWIVGNGHATDLWQNCSTSSSGNVHHCFSSSPNEWLQSVQATMILSIIFSILSLFLFFCQLFTLTKGGRFYITGIFQILAGLCVMSAAAIYTVRHPEWHLNSDYSYGFAYILAWVAFPLALLSGVIYVILRKRE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CMT1A/CMT1E/DSS/GAS-3/HMSNIA/HNPP/MGC20769/Sp110
Common Name PMP22
Gene Symbol PMP22
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.