Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PMS1 Partial ORF (NP_000525, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-439 10 ug
89-957-440 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-439 Supplier Abnova™ Supplier No. H00005378Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome. [provided by RefSeq]

Sequence: KELIENSLDAGATSVDVKLENYGFDKIEVRDNGEGIKAVDAPVMAMKYYTSKINSHEDLENLTTYGFRGEALGSICCIAEVLITTRTAADNFSTQYVLDGSGHILSQKPS

Specifications

Accession Number NP_000525
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5378
Molecular Weight (g/mol) 37.84kDa
Name PMS1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen KELIENSLDAGATSVDVKLENYGFDKIEVRDNGEGIKAVDAPVMAMKYYTSKINSHEDLENLTTYGFRGEALGSICCIAEVLITTRTAADNFSTQYVLDGSGHILSQKPS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp781M0253/FLJ98259/HNPCC3/PMSL1/hPMS1
Common Name PMS1
Gene Symbol PMS1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.