Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PNPT1 Full-length ORF (NP_149100.1, 1 a.a. - 783 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957508 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-508 10 μg
89-957-509 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-508 Supplier Abnova™ Supplier No. H00087178P01S

Used for AP, Array, ELISA, WB-Re

PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation (Raijmakers et al., 2002 [PubMed 12419256]).[supplied by OMIM]

Sequence: MAACRYCCSCLRLRPLSDGPFLLPRRDRALTQLQVRALWSSAGSRAVAVDLGNRKLEISSGKLARFADGSAVVQSGDTAVMVTAVSKTKPSPSQFMPLVVDYRQKAAAAGRIPTNYLRREVGTSDKEILTSRIIDRSIRPLFPAGYFYDTQVLCNLLAVDGVNEPDVLAINGASVALSLSDIPWNGPVGAVRIGIIDGEYVVNPTRKEMSSSTLNLVVAGAPKSQIVMLEASAENILQQDFCHAIKVGVKYTQQIIQGIQQLVKETGVTKRTPQKLFTPSPEIVKYTHKLAMERLYAVFTDYEHDKVSRDEAVNKIRLDTEEQLKEKFPEADPYEIIESFNVVAKEVFRSIVLNEYKRCDGRDLTSLRNVSCEVDMFKTLHGSALFQRGQTQVLCTVTFDSLESGIKSDQVITAINGIKDKNFMLHYEFPPYATNEIGKVTGLNRRELGHGALAEKALYPVIPRDFPFTIRVTSEVLESNGSSSMASACGGSLALMDSGVPISSAVAGVAIGLVTKTDPEKGEIEDYRLLTDILGIEDYNGDMDFKIAGTNKGITALQADIKLPGIPIKIVMEAIQQASVAKKEILQIMNKTISKPRASRKENGPVVETVQVPLSKRAKFVGPGGYNLKKLQAETGVTISQVDEETFSVFAPTPSAMHEARDFITEICKDDQEQQLEFGAVYTATITEIRDTGVMVKLYPNMTAVLLHNTQLDQRKIKHPTALGLEVGQEIQVKYFGRDPADGRMRLSRKVLQSPATTVVRTLNDRSSIVMGEPISQSSSNSQ

Specifications

Accession Number NP_149100.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 87178
Molecular Weight (g/mol) 112.3kDa
Name PNPT1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MAACRYCCSCLRLRPLSDGPFLLPRRDRALTQLQVRALWSSAGSRAVAVDLGNRKLEISSGKLARFADGSAVVQSGDTAVMVTAVSKTKPSPSQFMPLVVDYRQKAAAAGRIPTNYLRREVGTSDKEILTSRIIDRSIRPLFPAGYFYDTQVLCNLLAVDGVNEPDVLAINGASVALSLSDIPWNGPVGAVRIGIIDGEYVVNPTRKEMSSSTLNLVVAGAPKSQIVMLEASAENILQQDFCHAIKVGVKYTQQIIQGIQQLVKETGVTKRTPQKLFTPSPEIVKYTHKLAMERLYAVFTDYEHDKVSRDEAVNKIRLDTEEQLKEKFPEADPYEIIESFNVVAKEVFRSIVLNEYKRCDGRDLTSLRNVSCEVDMFKTLHGSALFQRGQTQVLCTVTFDSLESGIKSDQVITAINGIKDKNFMLHYEFPPYATNEIGKVTGLNRRELGHGALAEKALYPVIPRDFPFTIRVTSEVLESNGSSSMASACGGSLALMDSGVPISSAVAGVAIGLVTKTDPEKGEIEDYRLLTDILGIEDYNGDMDFKIAGTNKGITALQADIKLPGIPIKIVMEAIQQASVAKKEILQIMNKTISKPRASRKENGPVVETVQVPLSKRAKFVGPGGYNLKKLQAETGVTISQVDEETFSVFAPTPSAMHEARDFITEICKDDQEQQLEFGAVYTATITEIRDTGVMVKLYPNMTAVLLHNTQLDQRKIKHPTALGLEVGQEIQVKYFGRDPADGRMRLSRKVLQSPATTVVRTLNDRSSIVMGEPISQSSSNSQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp762K1914/OLD35/PNPASE/old-35
Common Name PNPT1
Gene Symbol PNPT1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.