Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human POLD1 Full-length ORF (AAH08800.1, 1 a.a. - 1107 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957539 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-539 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-539 Supplier Abnova™ Supplier No. H00005424P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The DNA polymerase delta complex is involved in DNA replication and repair, and it consists of the proliferating cell nuclear antigen (PCNA; MIM 176740), the multisubunit replication factor C (see MIM 102579), and the 4 subunit polymerase complex: POLD1, POLD2 (MIM 600815), POLD3 (MIM 611415), and POLD4 (MIM 611525) (Liu and Warbrick, 2006 [PubMed 16934752]).[supplied by OMIM]

Sequence: MDGKRRPGPGPGVPPKRARGGLWDDDDAPRPSQFEEDLALMEEMEAEHRLQEQEEEELQSVLEGVADGQVPPSAIDPRWLRPTPPALDPQTEPLIFQQLEIDHYVGPAQPVPGGPPPSHGSVPVLRAFGVTDEGFSVCCHIHGFAPYFYTPAPPGFGPEHMGDLQRELNLAISRDSRGGRELTGPAVLAVELCSRESMFGYHGHGPSPFLRITVALPRLVAPARRLLEQGIRVAGLGTPSFAPYEANVDFEIRFMVDTDIVGCNWLELPAGKYALRLKEKATQCQLEADVLWSDVVSHPPEGPWQRIAPLRVLSFDIECAGRKGIFPEPERDPVIQICSLGLRWGEPEPFLRLALTLRPCAPILGAKVQSYEKEEDLLQAWSTFIRIMDPDVITGYNIQNFDLPYLISRAQTLKVQTFPFLGRVAGLCSNIRDSSFQSKQTGRRDTKVVSMVGRVQMDMLQVLLREYKLRSYTLNAVSFHFLGEQKEDVQHSIITDLQNGNDQTRRRLAVYCLKDAYLPLRLLERLMVLVNAVEMARVTGVPLSYLLSRGQQVKVVSQLLRQAMHEGLLMPVVKSEGGEDYTGATVIEPLKGYYDVPIATLDFSSLYPSIMMAHNLCYTTLLRPGTAQKLGLTEDQFIRTPTGDEFVKTSVRKGLLPQILENLLSARKRAKAELAKETDPLRRQVLDGRQLALKVSANSVYGFTGAQVGKLPCLEISQSVTGFGRQMIEKTKQLVESKYTVENGYSTSAKVVYGDTDSVMCRFGVSSVAEAMALGREAADWVSGHFPSPIRLEFEKVYFPYLLISKKRYAGLLFSSRPDAHDRMDCKGLEAVRRDNCPLVANLVTASLRRLLIDRDPEGAVAHAQDVISDLLCNRIDISQLVITKELTRAASDYAGKQAHVELAERMRKRDPGSAPSLGDRVPYVIISAAKGVAAYMKSEDPLFVLEHSLPIDTQYYLEQQLAKPLLRIFEPILGEGRAEAVLLRGDHTRCKTVLTGKVGGLLAFAKRRNCCIGCRTVLSHQGAVCEFCQPRESELYQKEVSHLNALEERFSRLWTQCQRCQGSLHEDVICTSRDCPIFYMRKKVRKDLEDQEQLLRRFGPPGPEAW

Specifications

Accession Number AAH08800.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5424
Molecular Weight (g/mol) 150kDa
Name POLD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MDGKRRPGPGPGVPPKRARGGLWDDDDAPRPSQFEEDLALMEEMEAEHRLQEQEEEELQSVLEGVADGQVPPSAIDPRWLRPTPPALDPQTEPLIFQQLEIDHYVGPAQPVPGGPPPSHGSVPVLRAFGVTDEGFSVCCHIHGFAPYFYTPAPPGFGPEHMGDLQRELNLAISRDSRGGRELTGPAVLAVELCSRESMFGYHGHGPSPFLRITVALPRLVAPARRLLEQGIRVAGLGTPSFAPYEANVDFEIRFMVDTDIVGCNWLELPAGKYALRLKEKATQCQLEADVLWSDVVSHPPEGPWQRIAPLRVLSFDIECAGRKGIFPEPERDPVIQICSLGLRWGEPEPFLRLALTLRPCAPILGAKVQSYEKEEDLLQAWSTFIRIMDPDVITGYNIQNFDLPYLISRAQTLKVQTFPFLGRVAGLCSNIRDSSFQSKQTGRRDTKVVSMVGRVQMDMLQVLLREYKLRSYTLNAVSFHFLGEQKEDVQHSIITDLQNGNDQTRRRLAVYCLKDAYLPLRLLERLMVLVNAVEMARVTGVPLSYLLSRGQQVKVVSQLLRQAMHEGLLMPVVKSEGGEDYTGATVIEPLKGYYDVPIATLDFSSLYPSIMMAHNLCYTTLLRPGTAQKLGLTEDQFIRTPTGDEFVKTSVRKGLLPQILENLLSARKRAKAELAKETDPLRRQVLDGRQLALKVSANSVYGFTGAQVGKLPCLEISQSVTGFGRQMIEKTKQLVESKYTVENGYSTSAKVVYGDTDSVMCRFGVSSVAEAMALGREAADWVSGHFPSPIRLEFEKVYFPYLLISKKRYAGLLFSSRPDAHDRMDCKGLEAVRRDNCPLVANLVTASLRRLLIDRDPEGAVAHAQDVISDLLCNRIDISQLVITKELTRAASDYAGKQAHVELAERMRKRDPGSAPSLGDRVPYVIISAAKGVAAYMKSEDPLFVLEHSLPIDTQYYLEQQLAKPLLRIFEPILGEGRAEAVLLRGDHTRCKTVLTGKVGGLLAFAKRRNCCIGCRTVLSHQGAVCEFCQPRESELYQKEVSHLNALEERFSRLWTQCQRCQGSLHEDVICTSRDCPIFYMRKKVRKDLEDQEQLLRRFGPPGPEAW
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CDC2/POLD
Common Name POLD1
Gene Symbol POLD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.