Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human POLR3D Full-length ORF (NP_001713.2, 1 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-608 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-608 Supplier Abnova™ Supplier No. H00000661P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene complements a temperature-sensitive mutant isolated from the BHK-21 Syrian hamster cell line. It leads to a block in progression through the G1 phase of the cell cycle at nonpermissive temperatures. [provided by RefSeq]

Sequence: MSEGNAAGEPSTPGGPRPLLTGARGLIGRRPAPPLTPGRLPSIRSRDLTLGGVKKKTFTPNIISRKIKEEPKEEVTVKKEKRERDRDRQREGHGRGRGRPEVIQSHSIFEQGPAEMMKKKGNWDKTVDVSDMGPSHIINIKKEKRETDEETKQILRMLEKDDFLDDPGLRNDTRNMPVQLPLAHSGWLFKEENDEPDVKPWLAGPKEEDMEVDIPAVKVKEEPRDEEEEAKMKAPPKAARKTPGLPKDVSVAELLRELSLTKEEELLFLQLPDTLPGQPPTQDIKPIKTEVQGEDGQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDHKHR

Specifications

Accession Number NP_001713.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 661
Molecular Weight (g/mol) 70.8kDa
Name POLR3D (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSEGNAAGEPSTPGGPRPLLTGARGLIGRRPAPPLTPGRLPSIRSRDLTLGGVKKKTFTPNIISRKIKEEPKEEVTVKKEKRERDRDRQREGHGRGRGRPEVIQSHSIFEQGPAEMMKKKGNWDKTVDVSDMGPSHIINIKKEKRETDEETKQILRMLEKDDFLDDPGLRNDTRNMPVQLPLAHSGWLFKEENDEPDVKPWLAGPKEEDMEVDIPAVKVKEEPRDEEEEAKMKAPPKAARKTPGLPKDVSVAELLRELSLTKEEELLFLQLPDTLPGQPPTQDIKPIKTEVQGEDGQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDHKHR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BN51T/RPC4/RPC53/TSBN51
Common Name POLR3D
Gene Symbol POLR3D
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.