Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human POLRMT Partial ORF (NP_005026, 1121 a.a. - 1230 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-622 10 ug
89-957-623 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-622 Supplier Abnova™ Supplier No. H00005442Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes. [provided by RefSeq]

Sequence: PNFIHSLDSSHMMLTALHCYRKGLTFVSVHDCYWTHAADVSVMNQVCREQFVRLHSEPILQDLSRFLVKRFCSEPQKILEASQLKETLQAVPKPGAFDLEQVKRSTYFFS

Specifications

Accession Number NP_005026
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5442
Molecular Weight (g/mol) 37.84kDa
Name POLRMT (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen PNFIHSLDSSHMMLTALHCYRKGLTFVSVHDCYWTHAADVSVMNQVCREQFVRLHSEPILQDLSRFLVKRFCSEPQKILEASQLKETLQAVPKPGAFDLEQVKRSTYFFS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APOLMT/MTRNAP/MTRPOL/h-mtRPOL
Common Name POLRMT
Gene Symbol POLRMT
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.