Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human POU3F4 Full-length ORF (AAI46552.1, 1 a.a. - 361 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957692 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-692 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-692 Supplier Abnova™ Supplier No. H00005456P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the POU-III class of neural transcription factors. This gene may play a role in the mediation of epigenetic signals which induce striatal neuron-precursor differentiation. Mutations have been associated with X chromosome-linked nonsyndromic mixed deafness. [provided by RefSeq]

Sequence: MATAASNPYSILSSTSLVHADSAGMQQGSPFRNPQKLLQSDYLQGVPSNGHPLGHHWVTSLSDGGPWSSTLATSPLDQQDVKPGREDLQLGAIIHHRSPHVAHHSPHTNHPNAWGASPAPNPSITSSGQPLNVYSQPGFTVSGMLEHGGLTPPPAAASAQSLHPVLREPPDHGELGSHHCQDHSDEETPTSDELEQFAKQFKQRRIKLGFTQADVGLALGTLYGNVFSQTTICRFEGLQLSFKNMCKLKPLLNKWLEEADSSTGSPTSIDKIAAQGRKRKKRTSIEVSVKGVLETHFLKCPKPAAQEISSLADSLQLEKEVVRVWFCNRRQKEKRMTPPGDQQPHEVYSHTVKTDTSCHDL

Specifications

Accession Number AAI46552.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5456
Molecular Weight (g/mol) 66.66kDa
Name POU3F4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MATAASNPYSILSSTSLVHADSAGMQQGSPFRNPQKLLQSDYLQGVPSNGHPLGHHWVTSLSDGGPWSSTLATSPLDQQDVKPGREDLQLGAIIHHRSPHVAHHSPHTNHPNAWGASPAPNPSITSSGQPLNVYSQPGFTVSGMLEHGGLTPPPAAASAQSLHPVLREPPDHGELGSHHCQDHSDEETPTSDELEQFAKQFKQRRIKLGFTQADVGLALGTLYGNVFSQTTICRFEGLQLSFKNMCKLKPLLNKWLEEADSSTGSPTSIDKIAAQGRKRKKRTSIEVSVKGVLETHFLKCPKPAAQEISSLADSLQLEKEVVRVWFCNRRQKEKRMTPPGDQQPHEVYSHTVKTDTSCHDL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BRAIN-4/BRN4/DFN3/DFNX2/OTF9
Common Name POU3F4
Gene Symbol POU3F4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.