Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PP14/Glycodelin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB842100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB842100 100 μg
MAB842SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB842100 Supplier R&D Systems Supplier No. MAB842100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP14/Glycodelin Monoclonal specifically detects PP14/Glycodelin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PP14/Glycodelin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 929718
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P09466
Gene Alias alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PAEP, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PP14/Glycodelin, Met19-Phe180, Accession # P09466
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5047
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.