Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPA1 (NP_066952, 1 a.a. - 289 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89957715 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-715 Supplier Abnova™ Supplier No. P4908
Add to Cart
Add to Cart

Used for SDS-PAGE

The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMGSHMSGFSTEERAAPFSLEYRVFLKNEKGQYISPFHDIPIYADKDVFHMVVEVPRWSNAKMEIATKDPLNPIKQDVKKGKLRYVANLFPYKGYIWNYGAIPQTWEDPGHNDKHTGCCGDNDPIDVCEIGSKVCARGEIIGVKVLGILAMIDEGETDWKVIAINVDDPDAANYNDINDVKRLKPGYLEATVDWFRRYKVPDGKPENEFAFNAEFKDKDFAIDIIKSTHDHWKALVTKKTNGKGISCMNTTLSESPFKCDPDAARAIVDALPPPCESACTVPTDVDKWFHHQKN

Specifications

Accession Number NP_066952
Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5464
Molecular Weight (g/mol) 35.2kDa
Name PPA1 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMGSHMSGFSTEERAAPFSLEYRVFLKNEKGQYISPFHDIPIYADKDVFHMVVEVPRWSNAKMEIATKDPLNPIKQDVKKGKLRYVANLFPYKGYIWNYGAIPQTWEDPGHNDKHTGCCGDNDPIDVCEIGSKVCARGEIIGVKVLGILAMIDEGETDWKVIAINVDDPDAANYNDINDVKRLKPGYLEATVDWFRRYKVPDGKPENEFAFNAEFKDKDFAIDIIKSTHDHWKALVTKKTNGKGISCMNTTLSESPFKCDPDAARAIVDALPPPCESACTVPTDVDKWFHHQKN
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias IOPPP/MGC111556/PP/PP1/SID6-8061
Common Name PPA1
Gene Symbol PPA1
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.