Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PPAR-gamma Recombinant Protein

Catalog No. PIRP75684
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75684 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75684 Supplier Invitrogen™ Supplier No. RP75684
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

RP-75684 contains purified recombinant human PPAR gamma protein with an N-terminal hexahistidine tag. RP-75684 may be used as a positive control in many immunological assays.

This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P37231
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 0.02 mM EDTA, 50% glycerol, 75 mM NaCl and no preservative; pH 8
Gene ID (Entrez) 5468
Name Human PPAR-gamma
pH Range 8
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias CIMT1; GLM1; HUMPPARG; NR1C3; nuclear hormone receptor; nuclear hormone receptor; PPARG; Nuclear receptor subfamily 1 group C member 3; nuclear receptor transcription factor; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferative activated receptor, gamma; peroxisome proliferative activated receptor, gamma-2; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 1; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activated receptor gamma-1; peroxisome proliferator activated receptor gamma-2; peroxisome proliferator activatived receptor gamma; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; peroxisome proliferator-activated receptor; peroxisome proliferator-activated receptor gamma; peroxisome proliferator-activated receptor gamma 1; peroxisome proliferator-activated receptor gamma 1 A; peroxisome proliferator-activated receptor gamma 1-a; peroxisome proliferator-activated receptor gamma 1 b; peroxisome proliferator-activated receptor gamma 1-b; peroxisome proliferator-activated receptor gamma 1 c; peroxisome proliferator-activated receptor gamma 1 d; peroxisome proliferator-activated receptor gamma 2; peroxisome proliferator-activated receptor-gamma; peroxisome proliferator-activated receptor-gamma 2; peroxisome proliferator-cctivated receptor gamma 1; PPAR gamma; PPAR gamma 1; PPAR gamma 3; Pparg; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2; PPARy; proliferator activated gamma; transcription factor
Common Name PPAR gamma
Gene Symbol PPARG
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met29-Tyr505 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.