Learn More
Indigo Biosciences Human Peroxisome Proliferator-Activated Receptor alpha (PPARa, NR1C1) Reporter Assay Kit, 1x96-well assays plate format
Supplier:  Indigo Biosciences IB00111
Encompass_Preferred
This is an all-inclusive cell-based luciferase reporter assay kit targeting the Human Peroxisome Proliferator-Activated Receptor Alpha. INDIGO's Human PPAR alpha reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the Human PPAR alpha. In addition to PPAR alpha Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against human PPAR alpha. This kit provides researchers with: Clear, Reproducible Results, Exceptional Cell Viability Post-Thaw. Consistent Results Lot to Lot. Kits must be stored at -80C. Do not store in liquid nitrogen. Note Reporter Cells cannot be refrozen or maintained in extended culture.
Catalog No. 50-223-1756
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.