Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPFIBP2 Partial ORF (NP_003612, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957757 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-757 25 μg
89-957-756 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-757 Supplier Abnova™ Supplier No. H00008495Q01L

Used for AP, Array, ELISA, WB-Re

PPFIBP2, or liprin-beta-2, belongs to the liprin-beta family. See liprin-beta-1 (MIM 603141) for background on liprins.[supplied by OMIM]

Sequence: MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHS

Specifications

Accession Number NP_003612
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8495
Molecular Weight (g/mol) 36.74kDa
Name PPFIBP2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MASDASHALEAALEQMDGIIAGTKTGADLSDGTCEPGLASPASYMNPFPVLHLIEDLRLALEMLELPQERAALLSQIPGPTAAYIKEWFEESLSQVNHHS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Cclp1/DKFZp781K06126/MGC42541
Common Name PPFIBP2
Gene Symbol PPFIBP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.