Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPIA (NP_066953, 1 a.a. - 165 a.a.) Full-length Recombinant Protein

Catalog No. 89957768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-768 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-768 Supplier Abnova™ Supplier No. P3460
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

This gene encodes a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. The encoded protein is a cyclosporin binding-protein and may play a role in cyclosporin A-mediated immunosuppression. The protein can also interact with several HIV proteins, including p55 gag, Vpr, and capsid protein, and has been shown to be necessary for the formation of infectious HIV virions. Multiple pseudogenes that map to different chromosomes have been reported. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMVNPTVFFDIAVDGEPLGRVSFELFADKVPKTAENFRALSTGEKGFGYKGSCFHRIIPGFMCQGGDFTRHNGTGGKSIYGEKFEDENFILKHTGPGILSMANAGPNTNGSQFFICTAKTEWLDGKHVVFGKVKEGMNIVEAMERFGSRNGKTSKKITIADCGQLE

Specifications

Accession Number NP_066953
Concentration 1 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5478
Molecular Weight (g/mol) 20kDa
Name PPIA (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMVNPTVFFDIAVDGEPLGRVSFELFADKVPKTAENFRALSTGEKGFGYKGSCFHRIIPGFMCQGGDFTRHNGTGGKSIYGEKFEDENFILKHTGPGILSMANAGPNTNGSQFFICTAKTEWLDGKHVVFGKVKEGMNIVEAMERFGSRNGKTSKKITIADCGQLE
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg of protein (determined by LAL method)
Gene Alias CYPA/CYPH/MGC117158/MGC12404/MGC23397
Common Name PPIA
Gene Symbol PPIA
Biological Activity Specific activity is > 270nmoles/min/μg, and is defined as the amount of enzyme that cleaves 1μmole of suc-AAFP-pNA per minute at 25°C in Tris-HCl pH 8.0 using chymotrypsin.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.