Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPIF Full-length ORF (NP_005720.1, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957788 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-788 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-788 Supplier Abnova™ Supplier No. H00010105P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death. [provided by RefSeq]

Sequence: MLALRCGSRWLGLLSVPRSVPLRLPAARACSKGSGDPSSSSSSGNPLVYLDVDANGKPLGRVVLELKADVVPKTAENFRALCTGEKGFGYKGSTFHRVIPSFMCQAGDFTNHNGTGGKSIYGSRFPDENFTLKHVGPGVLSMANAGPNTNGSQFFICTIKTDWLDGKHVVFGHVKEGMDVVKKIESFGSKSGRTSKKIVITDCGQLS

Specifications

Accession Number NP_005720.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10105
Molecular Weight (g/mol) 48.4kDa
Name PPIF (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLALRCGSRWLGLLSVPRSVPLRLPAARACSKGSGDPSSSSSSGNPLVYLDVDANGKPLGRVVLELKADVVPKTAENFRALCTGEKGFGYKGSTFHRVIPSFMCQAGDFTNHNGTGGKSIYGSRFPDENFTLKHVGPGVLSMANAGPNTNGSQFFICTIKTDWLDGKHVVFGHVKEGMDVVKKIESFGSKSGRTSKKIVITDCGQLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CYP3/Cyp-D/FLJ90798/MGC117207
Common Name PPIF
Gene Symbol PPIF
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.