Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPIL5 Partial ORF (NP_982292, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957807 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-807 25 μg
89-957-806 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-807 Supplier Abnova™ Supplier No. H00122769Q01L

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene contains a leucine-rich repeat (LRR). It specifically interacts with TNFRSF9/4-1BB, a member of the tumor necrosis factor receptor (TNFR) superfamily. Overexpression of this gene suppresses the activation of NF-kappa B induced by TNFRSF9 or TNF receptor-associated factor 2 (TRAF2), which suggests that this protein is a negative regulator of TNFRSF9-mediated signaling cascades. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MKLHCEVEVISRHLPALGLRNRGKGVRAVLSLCQQTSRSQPPVRAFLLISTLKDKRGTRYELRENIEQFFTKFVDEGKATVRLKEPPVDICLSKDSIWLS

Specifications

Accession Number NP_982292
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 122769
Molecular Weight (g/mol) 36.74kDa
Name PPIL5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MKLHCEVEVISRHLPALGLRNRGKGVRAVLSLCQQTSRSQPPVRAFLLISTLKDKRGTRYELRENIEQFFTKFVDEGKATVRLKEPPVDICLSKDSIWLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 4-1BBLRR/LRR-1/MGC20689
Common Name PPIL5
Gene Symbol PPIL5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.