Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPM1D Full-length ORF (NP_003611.1, 1 a.a. - 605 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957815 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-815 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-815 Supplier Abnova™ Supplier No. H00008493P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. The expression of this gene is induced in a p53-dependent manner in response to various environmental stresses. While being induced by tumor suppressor protein TP53/p53, this phosphatase negatively regulates the activity of p38 MAP kinase, MAPK/p38, through which it reduces the phosphorylation of p53, and in turn suppresses p53-mediated transcription and apoptosis. This phosphatase thus mediates a feedback regulation of p38-p53 signaling that contributes to growth inhibition and the suppression of stress induced apoptosis. This gene is located in a chromosomal region known to be amplified in breast cancer. The amplification of this gene has been detected in both breast cancer cell line and primary breast tumors, which suggests a role of this gene in cancer development. [provided by RefSeq]

Sequence: MAGLYSLGVSVFSDQGGRKYMEDVTQIVVEPEPTAEEKPSPRRSLSQPLPPRPSPAALPGGEVSGKGPAVAAREARDPLPDAGASPAPSRCCRRRSSVAFFAVCDGHGGREAAQFAREHLWGFIKKQKGFTSSEPAKVCAAIRKGFLACHLAMWKKLAEWPKTMTGLPSTSGTTASVVIIRGMKMYVAHVGDSGVVLGIQDDPKDDFVRAVEVTQDHKPELPKERERIEGLGGSVMNKSGVNRVVWKRPRLTHNGPVRRSTVIDQIPFLAVARALGDLWSYDFFSGEFVVSPEPDTSVHTLDPQKHKYIILGSDGLWNMIPPQDAISMCQDQEEKKYLMGEHGQSCAKMLVNRALGRWRQRMLRADNTSAIVICISPEVDNQGNFTNEDELYLNLTDSPSYNSQETCVMTPSPCSTPPVKSLEEDPWPRVNSKDHIPALVRSNAFSENFLEVSAEIARENVQGVVIPSKDPEPLEENCAKALTLRIHDSLNNSLPIGLVPTNSTNTVMDQKNLKMSTPGQMKAQEIERTPPTNFKRTLEESNSGPLMKKHRRNGLSRSSGAQPASLPTTSQRKNSVKLTMRRRLRGQKKIGNPLLHQHRKTVCVC

Specifications

Accession Number NP_003611.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8493
Molecular Weight (g/mol) 93.1kDa
Name PPM1D (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Immunogen MAGLYSLGVSVFSDQGGRKYMEDVTQIVVEPEPTAEEKPSPRRSLSQPLPPRPSPAALPGGEVSGKGPAVAAREARDPLPDAGASPAPSRCCRRRSSVAFFAVCDGHGGREAAQFAREHLWGFIKKQKGFTSSEPAKVCAAIRKGFLACHLAMWKKLAEWPKTMTGLPSTSGTTASVVIIRGMKMYVAHVGDSGVVLGIQDDPKDDFVRAVEVTQDHKPELPKERERIEGLGGSVMNKSGVNRVVWKRPRLTHNGPVRRSTVIDQIPFLAVARALGDLWSYDFFSGEFVVSPEPDTSVHTLDPQKHKYIILGSDGLWNMIPPQDAISMCQDQEEKKYLMGEHGQSCAKMLVNRALGRWRQRMLRADNTSAIVICISPEVDNQGNFTNEDELYLNLTDSPSYNSQETCVMTPSPCSTPPVKSLEEDPWPRVNSKDHIPALVRSNAFSENFLEVSAEIARENVQGVVIPSKDPEPLEENCAKALTLRIHDSLNNSLPIGLVPTNSTNTVMDQKNLKMSTPGQMKAQEIERTPPTNFKRTLEESNSGPLMKKHRRNGLSRSSGAQPASLPTTSQRKNSVKLTMRRRLRGQKKIGNPLLHQHRKTVCVC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PP2C-DELTA/WIP1
Common Name PPM1D
Gene Symbol PPM1D
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.