Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP1R10 Full-length ORF (NP_002705.2, 1 a.a. - 940 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-852 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-852 Supplier Abnova™ Supplier No. H00005514P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6. [provided by RefSeq]

Sequence: MGSGPIDPKELLKGLDSFLNRDGEVKSVDGISKIFSLMKEARKMVSRCTYLNILLQTRSPEILVKFIDVGGYKLLNNWLTYSKTTNNIPLLQQILLTLQHLPLTVDHLKQNNTAKLVKQLSKSSEDEELRKLASVLVSDWMAVIRSQSSTQPAEKDKKKRKDEGKSRTTLPERPLTEVKAETRAEEAPEKKREKPKSLRTTAPSHAKFRSTGLELETPSLVPVKKNASTVVVSDKYNLKPIPLKRQSNVAAPGDATPPAEKKYKPLNTTPNATKEIKVKIIPPQPMEGLGFLDALNSAPVPGIKIKKKKKVLSPTAAKPSPFEGKTSTEPSTAKPSSPEPAPPSEAMDADRPGTPVPPVEVPELMDTASLEPGALDAKPVESPGDPNQLTRKGRKRKSVTWPEEGKLREYFYFELDETERVNVNKIKDFGEAAKREILSDRHAFETARRLSHDNMEEKVPWVCPRPLVLPSPLVTPGSNSQERYIQAEREKGILQELFLNKESPHEPDPEPYEPIPPKLIPLDEECSMDETPYVETLEPGGSGGSPDGAGGSKLPPVLANLMGSMGAGKGPQGPGGGGINVQEILTSIMGSPNSHPSEELLKQPDYSDKIKQMLVPHGLLGPGPIANGFPPGGPGGPKGMQHFPPGPGGPMPGPHGGPGGPVGPRLLGPPPPPRGGDPFWDGPGDPMRGGPMRGGPGPGPGPYHRGRGGRGGNEPPPPPPPFRGARGGRSGGGPPNGRGGPGGGMVGGGGHRPHEGPGGGMGNSSGHRPHEGPGGGMGSGHRPHEGPGGSMGGGGGHRPHEGPGGGISGGSGHRPHEGPGGGMGAGGGHRPHEGPGGSMGGSGGHRPHEGPGHGGPHGHRPHDVPGHRGHDHRGPPPHEHRGHDGPGHGGGGHRGHDGGHSHGGDMSNRPVCRHFMMKGNCRYENNCAFYHPGVNGPPLP

Specifications

Accession Number NP_002705.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5514
Molecular Weight (g/mol) 125.5kDa
Name PPP1R10 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MGSGPIDPKELLKGLDSFLNRDGEVKSVDGISKIFSLMKEARKMVSRCTYLNILLQTRSPEILVKFIDVGGYKLLNNWLTYSKTTNNIPLLQQILLTLQHLPLTVDHLKQNNTAKLVKQLSKSSEDEELRKLASVLVSDWMAVIRSQSSTQPAEKDKKKRKDEGKSRTTLPERPLTEVKAETRAEEAPEKKREKPKSLRTTAPSHAKFRSTGLELETPSLVPVKKNASTVVVSDKYNLKPIPLKRQSNVAAPGDATPPAEKKYKPLNTTPNATKEIKVKIIPPQPMEGLGFLDALNSAPVPGIKIKKKKKVLSPTAAKPSPFEGKTSTEPSTAKPSSPEPAPPSEAMDADRPGTPVPPVEVPELMDTASLEPGALDAKPVESPGDPNQLTRKGRKRKSVTWPEEGKLREYFYFELDETERVNVNKIKDFGEAAKREILSDRHAFETARRLSHDNMEEKVPWVCPRPLVLPSPLVTPGSNSQERYIQAEREKGILQELFLNKESPHEPDPEPYEPIPPKLIPLDEECSMDETPYVETLEPGGSGGSPDGAGGSKLPPVLANLMGSMGAGKGPQGPGGGGINVQEILTSIMGSPNSHPSEELLKQPDYSDKIKQMLVPHGLLGPGPIANGFPPGGPGGPKGMQHFPPGPGGPMPGPHGGPGGPVGPRLLGPPPPPRGGDPFWDGPGDPMRGGPMRGGPGPGPGPYHRGRGGRGGNEPPPPPPPFRGARGGRSGGGPPNGRGGPGGGMVGGGGHRPHEGPGGGMGNSSGHRPHEGPGGGMGSGHRPHEGPGGSMGGGGGHRPHEGPGGGISGGSGHRPHEGPGGGMGAGGGHRPHEGPGGSMGGSGGHRPHEGPGHGGPHGHRPHDVPGHRGHDHRGPPPHEHRGHDGPGHGGGGHRGHDGGHSHGGDMSNRPVCRHFMMKGNCRYENNCAFYHPGVNGPPLP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CAT53/FB19/PNUTS/PP1R10
Common Name PPP1R10
Gene Symbol PPP1R10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.