Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP1R12B Full-length ORF (ENSP00000349206, 1 a.a. - 386 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957854 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-854 25 ug
89-957-853 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-854 Supplier Abnova™ Supplier No. H00004660P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Myosin light chain phosphatase (MLCP) consists of three subunits- catalytic subunit, large subunit/myosin binding subunit (MBS) and small subunit (sm-M20). This gene is a multi-functional gene which encodes both MBS and sm-M20. MLCP regulates myosins and the dephosphorylation is enhanced by the presence of MBS. The sm-M20 is suggested to play a regulatory role in muscle contraction by binding to MBS. MBS is also encoded by another gene, myosin light chain phosphatase target subunit 1. sm-M20 shows higher binding affinity to this gene product than to myosin light chain phosphatase target subunit 2-MBS even though the two MBS proteins are highly similar. Although both MBSs increase the activity of MLCP, myosin light chain phosphatase target subunit 1-MBS is a more efficient activator. There are four alternatively spliced transcript variants described; two alter the MBS coding region and two alter the sm-M20 coding region of this gene. [provided by RefSeq]

Sequence: MAELEHLGGKRAESARMRRAEQLRRWRGSLTEQEPAERRGAGRQPLTRRGSPRVRFEDGAVFLAACSSGDTDEVRKLLARGADINTVNVDGLTALHQACIDENLDMVKFLVENRANVNQQDNEGWTPLHAAASCGYLNIAEYFINHGASVGIVNSEGEVPSDLAEEPAMKDLLLEQVKKQGVDLEQSRKEEEQQMLQDARQWLNSGKIEDVRQARSGATALHVAAAKGYSEVLRLLIQAGYELNVQDYDGWTPLHAAAHWGVKEACSILAEALCDMDIRNKLGQTPFDVADEGLVEHLELLQKKQNVLRSEKETRNKLIESDLNSKIQSGFFKNKEKMLYEEETPKSQEMEEENKESSSSSSEEEEGEDEASESETEKEAVLFWPF

Specifications

Accession Number ENSP00000349206
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4660
Molecular Weight (g/mol) 69.7kDa
Name PPP1R12B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MAELEHLGGKRAESARMRRAEQLRRWRGSLTEQEPAERRGAGRQPLTRRGSPRVRFEDGAVFLAACSSGDTDEVRKLLARGADINTVNVDGLTALHQACIDENLDMVKFLVENRANVNQQDNEGWTPLHAAASCGYLNIAEYFINHGASVGIVNSEGEVPSDLAEEPAMKDLLLEQVKKQGVDLEQSRKEEEQQMLQDARQWLNSGKIEDVRQARSGATALHVAAAKGYSEVLRLLIQAGYELNVQDYDGWTPLHAAAHWGVKEACSILAEALCDMDIRNKLGQTPFDVADEGLVEHLELLQKKQNVLRSEKETRNKLIESDLNSKIQSGFFKNKEKMLYEEETPKSQEMEEENKESSSSSSEEEEGEDEASESETEKEAVLFWPF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC131980/MGC87886/MYPT2
Common Name PPP1R12B
Gene Symbol PPP1R12B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.