Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP1R3A Partial ORF (NP_002702.1, 3 a.a. - 101 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957888 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-888 25 ug
89-957-887 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-888 Supplier Abnova™ Supplier No. H00005506Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The glycogen-associated form of protein phosphatase-1 (PP1) derived from skeletal muscle is a heterodimer composed of a 37-kD catalytic subunit and a 124-kD targeting and regulatory subunit. This gene encodes the regulatory subunit which binds to muscle glycogen with high affinity, thereby enhancing dephosphorylation of glycogen-bound substrates for PP1 such as glycogen synthase and glycogen phosphorylase kinase. [provided by RefSeq]

Sequence: PSEVPSQISKDNFLEVPNLSDSLCEDEEVTFQPGFSPQPSRRGSDSSEDIYLDTPSSGTRRVSFADSFGFNLVSVKEFDCWELPSASTTFDLGTDIFHT

Specifications

Accession Number NP_002702.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5506
Molecular Weight (g/mol) 36.63kDa
Name PPP1R3A (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen PSEVPSQISKDNFLEVPNLSDSLCEDEEVTFQPGFSPQPSRRGSDSSEDIYLDTPSSGTRRVSFADSFGFNLVSVKEFDCWELPSASTTFDLGTDIFHT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GM/PP1G/PPP1R3
Common Name PPP1R3A
Gene Symbol PPP1R3A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.