Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP2R1B Partial ORF (AAH27596, 500 a.a. - 599 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-911 25 ug
89-957-910 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-911 Supplier Abnova™ Supplier No. H00005519Q01L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

This gene encodes a constant regulatory subunit of protein phosphatase 2. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. This gene encodes a beta isoform of the constant regulatory subunit A. Defects in this gene could be the cause of some lung and colon cancers. At least two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: VMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVVA

Specifications

Accession Number AAH27596
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5519
Molecular Weight (g/mol) 36.63kDa
Name PPP2R1B (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen VMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVVA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC26454/PR65B
Common Name PPP2R1B
Gene Symbol PPP2R1B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.