Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP2R5B Full-length ORF (NP_006235.1, 1 a.a. - 497 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957942 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-942 25 ug
89-957-941 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-942 Supplier Abnova™ Supplier No. H00005526P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B56 subfamily. [provided by RefSeq]

Sequence: METKLPPASTPTSPSSPGLSPVPPPDKVDGFSRRSLRRARPRRSHSSSQFRYQSNQQELTPLPLLKDVPASELHELLSRKLAQCGVMFDFLDCVADLKGKEVKRAALNELVECVGSTRGVLIEPVYPDIIRMISVNIFRTLPPSENPEFDPEEDEPNLEPSWPHLQLVYEFFLRFLESPDFQPSVAKRYVDQKFVLMLLELFDSEDPREREYLKTILHRVYGKFLGLRAYIRKQCNHIFLRFIYEFEHFNGVAELLEILGSIINGFALPLKTEHKQFLVRVLIPLHSVKSLSVFHAQLAYCVVQFLEKDATLTEHVIRGLLKYWPKTCTQKEVMFLGEMEEILDVIEPSQFVKIQEPLFKQVARCVSSPHFQVAERALYFWNNEYILSLIEDNCHTVLPAVFGTLYQVSKEHWNQTIVSLIYNVLKTFMEMNGKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTPQVAASGGQS

Specifications

Accession Number NP_006235.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5526
Molecular Weight (g/mol) 83.8kDa
Name PPP2R5B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen METKLPPASTPTSPSSPGLSPVPPPDKVDGFSRRSLRRARPRRSHSSSQFRYQSNQQELTPLPLLKDVPASELHELLSRKLAQCGVMFDFLDCVADLKGKEVKRAALNELVECVGSTRGVLIEPVYPDIIRMISVNIFRTLPPSENPEFDPEEDEPNLEPSWPHLQLVYEFFLRFLESPDFQPSVAKRYVDQKFVLMLLELFDSEDPREREYLKTILHRVYGKFLGLRAYIRKQCNHIFLRFIYEFEHFNGVAELLEILGSIINGFALPLKTEHKQFLVRVLIPLHSVKSLSVFHAQLAYCVVQFLEKDATLTEHVIRGLLKYWPKTCTQKEVMFLGEMEEILDVIEPSQFVKIQEPLFKQVARCVSSPHFQVAERALYFWNNEYILSLIEDNCHTVLPAVFGTLYQVSKEHWNQTIVSLIYNVLKTFMEMNGKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTPQVAASGGQS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias B56B/FLJ35411/PR61B
Common Name PPP2R5B
Gene Symbol PPP2R5B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.