Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP2R5E Full-length ORF (NP_006237.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957952 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-952 10 ug
89-957-953 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-952 Supplier Abnova™ Supplier No. H00005529P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an epsilon isoform of the regulatory subunit B56 subfamily. [provided by RefSeq]

Sequence: MSSAPTTPPSVDKVDGFSRKSVRKARQKRSQSSSQFRSQGKPIELTPLPLLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPHLQLVYEFFIRFLESQEFQPSIAKKYIDQKFVLQLLELFDSEDPRERDYLKTVLHRIYGKFLGLRAFIRKQINNIFLRFVYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLVKVLIPLHTVRSLSLFHAQLAYCIVQFLEKDPSLTEPVIRGLMKFWPKTCSQKEVMFLGELEEILDVIEPSQFVKIQEPLFKQIAKCVSSPHFQVAERALYYWNNEYIMSLIEENSNVILPIMFSSLYRISKEHWNPAIVALVYNVLKAFMEMNSTMFDELTATYKSDRQREKKKEKEREELWKKLEDLELKRGLRRDGIIPT

Specifications

Accession Number NP_006237.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5529
Molecular Weight (g/mol) 81.1kDa
Name PPP2R5E (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MSSAPTTPPSVDKVDGFSRKSVRKARQKRSQSSSQFRSQGKPIELTPLPLLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPHLQLVYEFFIRFLESQEFQPSIAKKYIDQKFVLQLLELFDSEDPRERDYLKTVLHRIYGKFLGLRAFIRKQINNIFLRFVYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLVKVLIPLHTVRSLSLFHAQLAYCIVQFLEKDPSLTEPVIRGLMKFWPKTCSQKEVMFLGELEEILDVIEPSQFVKIQEPLFKQIAKCVSSPHFQVAERALYYWNNEYIMSLIEENSNVILPIMFSSLYRISKEHWNPAIVALVYNVLKAFMEMNSTMFDELTATYKSDRQREKKKEKEREELWKKLEDLELKRGLRRDGIIPT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name PPP2R5E
Gene Symbol PPP2R5E
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.