Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPP3CC Partial ORF (NP_005596.2, 1 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957961 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-961 10 ug
89-957-962 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-961 Supplier Abnova™ Supplier No. H00005533Q01S

Used for AP, Array, ELISA, WB-Re

Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca(2+)-dependent modifier of phosphorylation status. In testis, the motility of the sperm is thought to be controlled by cAMP-dependent phosphorylation and a unique form of calcineurin appears to be associated with the flagellum. The calcineurin holoenzyme is composed of catalytic and regulatory subunits of 60 and 18 kD, respectively. At least 3 genes, calcineurin A-alpha (CALNA1; MIM 114105), calcineurin A-beta (CALNA2; MIM 114106), and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).[supplied by OMIM]

Sequence: MSGRRFHLSTTDRVIKAVPFPPTQRLTFKEVFENGKPKVDVLKNHLVKEGRLEEEVALKIINDGAAILRQEKTMIEVDAPI

Specifications

Accession Number NP_005596.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5533
Molecular Weight (g/mol) 34.65kDa
Name PPP3CC (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MSGRRFHLSTTDRVIKAVPFPPTQRLTFKEVFENGKPKVDVLKNHLVKEGRLEEEVALKIINDGAAILRQEKTMIEVDAPI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CALNA3
Common Name PPP3CC
Gene Symbol PPP3CC
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.