Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPT1 Full-length ORF (NP_000301.1, 1 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957979 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-979 10 ug
89-957-980 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-979 Supplier Abnova™ Supplier No. H00005538P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene

Sequence: MASPGCLWLLAVALLPWTCASRALQHLDPPAPLPLVIWHGMGDSCCNPLSMGAIKKMVEKKIPGIYVLSLEIGKTLMEDVENSFFLNVNSQVTTVCQALAKDPKLQQGYNAMGFSQGGQFLRAVAQRCPSPPMINLISVGGQHQGVFGLPRCPGESSHICDFIRKTLNAGAYSKVVQERLVQAEYWHDPIKEDVYRNHSIFLADINQERGINESYKKNLMALKKFVMVKFLNDSIVDPVDSEWFGFYRSGQAKETIPLQETSLYTQDRLGLKEMDNAGQLVFLATEGDHLQLSEEWFYAHIIPFLG

Specifications

Accession Number NP_000301.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5538
Molecular Weight (g/mol) 60.6kDa
Name PPT1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MASPGCLWLLAVALLPWTCASRALQHLDPPAPLPLVIWHGMGDSCCNPLSMGAIKKMVEKKIPGIYVLSLEIGKTLMEDVENSFFLNVNSQVTTVCQALAKDPKLQQGYNAMGFSQGGQFLRAVAQRCPSPPMINLISVGGQHQGVFGLPRCPGESSHICDFIRKTLNAGAYSKVVQERLVQAEYWHDPIKEDVYRNHSIFLADINQERGINESYKKNLMALKKFVMVKFLNDSIVDPVDSEWFGFYRSGQAKETIPLQETSLYTQDRLGLKEMDNAGQLVFLATEGDHLQLSEEWFYAHIIPFLG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CLN1/INCL/PPT
Common Name PPT1
Gene Symbol PPT1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.