Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PPT2 Full-length ORF (NP_005146.3, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-984 25 μg
89-957-983 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-984 Supplier Abnova™ Supplier No. H00009374P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the palmitoyl-protein thioesterase family. The encoded glycosylated lysosomal protein has palmitoyl-CoA hydrolase activity in vitro, but does not hydrolyze palmitate from cysteine residues in proteins. Three transcript variants encoding different isoforms have been described for this gene, with one of the isoforms being inactive. [provided by RefSeq]

Sequence: MLGLWGQRLPAAWVLLLLPFLPLLLLAAPAPHRASYKPVIVVHGLFDSSYSFRHLLEYINETHPGTVVTVLDLFDGRESLRPLWEQVQGFREAVVPIMAKAPQGVHLICYSQGGLVCRALLSVMDDHNVDSFISLSSPQMGQYGDTDYLKWLFPTSMRSNLYRICYSPWGQEFSICNYWHDPHHDDLYLNASSFLALINGERDHPNATVWRKNFLRVGHLVLIGGPDDGVITPWQSSFFGFYDANETVLEMEEQLVYLRDSFGLKTLLARGAIVRCPMAGISHTAWHSNRTLYETCIEPWLS

Specifications

Accession Number NP_005146.3
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9374
Molecular Weight (g/mol) 60.7kDa
Name PPT2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLGLWGQRLPAAWVLLLLPFLPLLLLAAPAPHRASYKPVIVVHGLFDSSYSFRHLLEYINETHPGTVVTVLDLFDGRESLRPLWEQVQGFREAVVPIMAKAPQGVHLICYSQGGLVCRALLSVMDDHNVDSFISLSSPQMGQYGDTDYLKWLFPTSMRSNLYRICYSPWGQEFSICNYWHDPHHDDLYLNASSFLALINGERDHPNATVWRKNFLRVGHLVLIGGPDDGVITPWQSSFFGFYDANETVLEMEEQLVYLRDSFGLKTLLARGAIVRCPMAGISHTAWHSNRTLYETCIEPWLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C6orf8/DKFZp564P1516/G14
Common Name PPT2
Gene Symbol PPT2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.