Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRCP Full-length ORF (NP_005031.1, 1 a.a. - 496 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958016 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-016 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-016 Supplier Abnova™ Supplier No. H00005547P02
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a lysosomal prolylcarboxypeptidase, which cleaves C-terminal amino acids linked to proline in peptides such as angiotension II, III and des-Arg9-bradykinin. The cleavage occurs at acidic pH, but the enzyme activity is retained with some substrates at neutral pH. This enzyme has been shown to be an activator of the cell matrix-associated prekallikrein. The importance of angiotension II, one of the substrates of this enzyme, in regulating blood pressure and electrolyte balance suggests that this gene may be related to essential hypertension. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MGRRALLLLLLSFLAPWATIALRPALRALGSLHLPTNPTSLPAVAKNYSVLYFQQKVDHFGFNTVKTFNQRYLVADKYWKKNGGSILFYTGNEGDIIWFCNNTGFMWDVAEELKAMLVFAEHRYYGESLPFGDNSFKDSRHLNFLTSEQALADFAELIKHLKRTIPGAENQPVIAIGGSYGGMLAAWFRMKYPHMVVGALAASAPIWQFEDLVPCGVFMKIVTTDFRKSGPHCSESIHRSWDAINRLSNTGSGLQWLTGALHLCSPLTSQDIQHLKDWISETWVNLAMVDYPYASNFLQPLPAWPIKVVCQYLKNPNVSDSLLLQNIFQALNVYYNYSGQVKCLNISETATSSLGTLGWSYQACTEVVMPFCTNGVDDMFEPHSWNLKELSDDCFQQWGVRPRPSWITTMYGGKNISSHTNIVFSNGELDPWSGGGVTKDITDTLVAVTISEGAHHLDLRTKNALDPMSVLLARSLEVRHMKNWIRDFYDSAGKQH

Specifications

Accession Number NP_005031.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5547
Molecular Weight (g/mol) 82.2kDa
Name PRCP (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MGRRALLLLLLSFLAPWATIALRPALRALGSLHLPTNPTSLPAVAKNYSVLYFQQKVDHFGFNTVKTFNQRYLVADKYWKKNGGSILFYTGNEGDIIWFCNNTGFMWDVAEELKAMLVFAEHRYYGESLPFGDNSFKDSRHLNFLTSEQALADFAELIKHLKRTIPGAENQPVIAIGGSYGGMLAAWFRMKYPHMVVGALAASAPIWQFEDLVPCGVFMKIVTTDFRKSGPHCSESIHRSWDAINRLSNTGSGLQWLTGALHLCSPLTSQDIQHLKDWISETWVNLAMVDYPYASNFLQPLPAWPIKVVCQYLKNPNVSDSLLLQNIFQALNVYYNYSGQVKCLNISETATSSLGTLGWSYQACTEVVMPFCTNGVDDMFEPHSWNLKELSDDCFQQWGVRPRPSWITTMYGGKNISSHTNIVFSNGELDPWSGGGVTKDITDTLVAVTISEGAHHLDLRTKNALDPMSVLLARSLEVRHMKNWIRDFYDSAGKQH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HUMPCP/MGC2202/PCP
Common Name PRCP
Gene Symbol PRCP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.