Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRDM2 Partial ORF (NP_036363, 1606 a.a. - 1705 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958025 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-025 25 μg
89-958-024 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-958-025 Supplier Abnova™ Supplier No. H00007799Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This tumor suppressor gene is a member of a nuclear histone/protein methyltransferase superfamily. It encodes a zinc finger protein that can bind to retinoblastoma protein, estrogen receptor, and the TPA-responsive element (MTE) of the heme-oxygenase-1 gene. Although the functions of this protein have not been fully characterized, it may (1) play a role in transcriptional regulation during neuronal differentiation and pathogenesis of retinoblastoma, (2) act as a transcriptional activator of the heme-oxygenase-1 gene, and (3) be a specific effector of estrogen action. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KTSRSLHVRVQKSKAVLQSKSTLASKKRTDRFNIKSRERSGGPVTRSLQLAAAADLSENKREDGSAKQELKDFSYSLRLASRCSPPAAPYITRQYRKVKA

Specifications

Accession Number NP_036363
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7799
Molecular Weight (g/mol) 36.74kDa
Name PRDM2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen KTSRSLHVRVQKSKAVLQSKSTLASKKRTDRFNIKSRERSGGPVTRSLQLAAAADLSENKREDGSAKQELKDFSYSLRLASRCSPPAAPYITRQYRKVKA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HUMHOXY1/KMT8/MTB-ZF/RIZ/RIZ1/RIZ2
Common Name PRDM2
Gene Symbol PRDM2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.