Learn More
Sino Biological Human PRDX6 MAb 25TESTS 11257-MM03-F-25
Supplier:  Sino Biological 11257MM03F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Peroxiredoxin 6 / PRDX6 (rh Peroxiredoxin 6 / PRDX6; Catalog#11257-H07E; P30041; Met1-Pro224) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-9587
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.