Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate Ig Lambda Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7235380
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB100492SP 25 μg
MAB10049210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB100492SP Supplier R&D Systems Supplier No. MAB100492SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Human lambda light chain Monoclonal specifically detects Human lambda light chain in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen Human lambda light chain
Applications Western Blot
Classification Monoclonal
Clone 2496A
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Alias Bence Jones protein, BJP, Constant region of lambda light chains, HCAK 1, HCAK1, Ig kappa chain C region, Ig lambda chain C regions, IGKC, IGLC, IGLC 1, IGLC 2, IGLC 3, IGLC1, IGLC2, IGLC3, Immunoglobulin kappa constant, Immunoglobulin kappa constant region, Immunoglobulin kappa light chain, Immunoglobulin lambda constant 1, Kappa 1 immunoglobulin light chain, Km, Mcg marker, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809, Paraprotein
Host Species Rabbit
Immunogen Human Ig Lamda Light Chain
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.